Learn More
MP Biomedicals™ Yeast Extract Peptone Dextrose Agar
Product Code. 11317759 Shop All MP Biomedicals Products
Change view
Click to view available options
Concentration:
67 g/L
Unit Size:
454g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Concentration unitSize
11317759 67 g/L 454g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11317759 Supplier MP Biomedicals™ Supplier No. 114001222
MP Biomedicals™ YPD Agar is used for routine growth of Saccharomyces cerevisiae for molecular biology studies.
  • YPD (YEPD) agar is a nutritionally rich medium for the growth of S. cerevisiae containing dextrose, peptone, yeast extract and agar.
  • Dextrose is the energy source.
  • Yeast extract and peptone provide carbon, nitrogen, minerals, vitamins, trace ingredients and other essential growth nutrients which stimulate the growth of yeasts.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 67 g/L
Form Powder
Grade Molecular Biology Certified
For Use With (Application) Growth of Saccharomyces cerevisiae
pH 6.2 ±0.3 (at 25°C)
Weight (Metric) 454 g
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Politique de confidentialité.