missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ YARS Recombinant Protein

Brand:  Abnova™ H00008565-P01.2ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192592

  • 2815.00 NOK / 2µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

Antibody Production, ELISA, Protein Array, Western Blot
83.82
8
Glutathione Sepharose 4 Fast Flow
2μg
-80°C
Recombinant
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Human YARS Full-length ORF Recombinant Protein with GST-tag at N-terminal
In vitro wheat germ expression system
12.5% SDS-PAGE stained with Coomassie Blue
Wheat Germ (in vitro)
Human
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers