missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YAP1 Monoclonal Antibody (OTI6B11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813311S
This item is not returnable.
View return policy
Description
YAP1 Monoclonal antibody specifically detects YAP1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| YAP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P46937, P46938 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 10413, 22601 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI6B11 | |
| Unconjugated | |
| YAP1 | |
| 65 kDa Yes-associated protein; AI325207; cb194; CH211-181P1.5-001; COB1; Protein yorkie homolog; sb:cb194; si:ch211-181p1.5; si:dkey-3b8.3; transcriptional coactivator YAP1; wu:fc18c04; yap; Yap1; YAP2; Yap65; Yes associated protein 1; Yes1 associated transcriptional regulator; yes-associated protein 1; yes-associated protein 2; yes-associated protein YAP65 homolog; yes-associated protein, 65 kDa; Yki; Yorkie; yorkie homolog; zgc:158380; zYAP | |
| Human recombinant protein fragment corresponding to amino acids 347-504 of human YAP1 (NP_001123617) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction