missing translation for 'onlineSavingsMsg'
Learn More

Yangonin-d3, TRC

Product Code. 30453524
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453524 25 mg 25mg
30456281 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30453524 Supplier Toronto Research Chemicals Supplier No. TRCY10055225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C15H11D3O4
Molecular Weight (g/mol) 261.29
Synonym 4-Methoxy-6-[(1E)-2-(4-methoxyphenyl)ethenyl]-2H-pyran-2-one-d3, 4-Methoxy-6-[β-(p-anisyl)vinyl]-α-pyrone-d3, 6-(p-Methoxystyryl)-4-methoxy-α-pyrone-d3
SMILES O=C1C=C(OC([2H])([2H])[2H])C=C(/C=C/C2=CC=C(OC)C=C2)O1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H14O4/c1-17-12-6-3-11(4-7-12)5-8-13-9-14(18-2)10-15(16)19-13/h3-10H,1-2H3/b8-5+/i2D3
Chemical Name or Material Yangonin-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.