Learn More
Xylometazoline hydrochloride, MedChemExpress
Product Code. 16492189
Encompass_Preferred
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. 16492189
Brand: Medchem Express HYB0475
MedChemExpress Xylometazoline hydrochloride is an α-adrenoceptor agonist commonly used as nasal decongestant.
Xylometazoline hydrochloride is an α-adrenoceptor agonist commonly used as nasal decongestant.
Specifications
Chemical Name or Material Xylometazoline hydrochloride
Physical Form Solid
CAS 1218-35-5
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C16H25ClN2
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 50 mg/mL (178.04 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (89.02 mM; Need ultrasonic)
SMILES CC1=C(CC2=NCCN2)C(C)=CC(C(C)(C)C)=C1.Cl
Molecular Weight (g/mol) 280.84
Formula Weight 280.84
Percent Purity 99.58%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.