Learn More
Scharlab XYLEN PSS 1L
Brand:  Scharlab XI00501000
XYLEN PSS 1L
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11352635
  • ONLINE EXCLUSIVE PRICE
    402.15 NOK
    350.51 NOK / 1 L
    Save 51.64 NOK
    12% Off