Learn More
Honeywell™ XV100 Series Safety Glasses
Description
- 10-base, wrap-around, uni-lens for great peripheral vision and protection
- Molded, universal nosepiece for easy fit
- Lightweight design and thin, shaped temples for all-day comfort
- Protects against impact (45 m/s)
- Well suited for use in light and heavy manufacturing, utilities, biotech, ship building, construction, and automotive manufacturing
Specifications
Specifications
| Certifications/Compliance | EC Category PPE: 1, In conformity with Council Directive 89/686/EEC and associated amendments, NIOSH 1979 |
| Lens Coating | Scratch-resistant |
| Description | Model XV101 |
| Certifications/Compliance (Europe) | EC Category PPE: 1, In conformity with Council Directive 89/686/EEC and associated amendments, NIOSH 1979 |
| Lens Tint | Gray |
| For Use With (Application) | General use and protection, utilities, light and heavy manufacturing, biotech, ship building, construction, automotive manufacturing |
| Frame Color | Gray |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |