Learn More
xSodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl diphosphate, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 20398-34-9 |
| Molecular Formula | C10H15N5O10P2.xNa |
| MDL Number | MFCD00065987 |
| Quantity | 1 g |
| SMILES | O=P(OP(O)([O-])=O)(OC[C@H]1O[C@H]([C@@H]([C@@H]1O)O)N2C=NC3=C(N=CN=C23)N)O.[Na+] |
| Percent Purity | 95% |
| Chemical Name or Material | xSodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl diphosphate |