Learn More
XPHOS, TRC
Specifications
Specifications
| Chemical Name or Material | XPHOS |
| CAS | 564483-18-7 |
| Molecular Formula | C33 H49 P |
| Quantity | 500 mg |
| Synonym | Phosphine','dicyclohexyl[2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl]-, Dicyclohexyl[2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl]phosphine (ACI), 2,,4',6'-Diisopropyl-1,1'-biphenyl-2-yldicyclohexylphosphine, 2-(Dicyclohexylphosphino)-2',4',6'-tri(isopropyl)biphenyl, 2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl, 2-Dicyclohexylphosphino-2',4',6'-triisoprophylbiphenyl, [2',4',6'-Triisopropyl-1,1'-biphenyl-2-yl]dicyclohexylphosphine, Dicyclohexyl(2',4',6'-triisopropyl[1,1'-biphenyl]-2-yl)phosphine, Dicyclohexyl(2',4',6'-triisopropylbiphenyl-2-yl)phosphine, X-Phos |
| InChI Formula | InChI=1S/C33H49P/c1-23(2)26-21-30(24(3)4)33(31(22-26)25(5)6)29-19-13-14-20-32(29)34(27-15-9-7-10-16-27)28-17-11-8-12-18-28/h13-14,19-25,27-28H,7-12,15-18H2,1-6H3 |
| SMILES | CC(C)c1cc(C(C)C)c(c(c1)C(C)C)c2ccccc2P(C3CCCCC3)C4CCCCC4 |
| IUPAC Name | dicyclohexyl-[2-[2,4,6-tri(propan-2-yl)phenyl]phenyl]phosphane |
| Molecular Weight (g/mol) | 476.72 |
| Formula Weight | 476.3572 |