missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
XPE/DDB2 Polyclonal antibody specifically detects XPE/DDB2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | XPE/DDB2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1:200-1:1000 |
| Formulation | 0.2 μm filtered solution in PBS |
| Gene Alias | damage-specific DNA binding protein 2 (48kD), damage-specific DNA binding protein 2, 48kDa, Damage-specific DNA-binding protein 2, DDB p48 subunit, DDBb, DNA damage-binding protein 2, FLJ34321, UV-damaged DNA-binding protein 2, UV-DDB 2, UV-DDB2, xeroderma pigmentosum group E protein |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human XPE/DDB2. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?