Learn More
Invitrogen™ XPD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120282
Description
Positive test controls include: HeLa, K-562, 293T, MCF7, mouse lung, mouse testis, rat testis. The target is usually found in the following locations: Cytoplasm, Nucleus, cytoskeleton, spindle. Immunogen sequence: MRELKRTLDA KGHGVLEMPS GTGKTVSLLA LIMAYQRAYP LEVTKLIYCS RTVPEIEKVI EELRKLLNFY EKQEGEKLPF LGLALSSRKN LCIHPEVTPL RFGKDVDGKC HSLTASYVRA QYQHDTSLPH CRFYEEFDAH GREVPLPAGI YNLDDLKALG RRQGWCPYFL ARYSILHANV.
XPD is involved with the nucleotide excision repair pathway that is a mechanism to repair damage to DNA. The protein encoded by this gene is involved in transcription-coupled nucleotide excision repair and is an integral member of the basal transcription factor BTF2/TFIIH complex. The gene product has ATP-dependent DNA helicase activity and belongs to the RAD3/XPD subfamily of helicases. Defects in this gene can result in three different disorders, the cancer-prone syndrome xeroderma pigmentosum complementation group D, trichothiodystrophy, and Cockayne syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| XPD | |
| Polyclonal | |
| Unconjugated | |
| ERCC2 | |
| AA407812; AU020867; AW240756; basic transcription factor 2 80 kDa subunit; BTF2 p80; COFS2; CXPD; DNA excision repair protein ERCC-2; DNA repair protein complementing XP-D cells; EM9; ERCC excision repair 2, TFIIH core complex helicase subunit; Ercc2; Ercc-2; excision repair cross-complementation group 2; excision repair cross-complementing rodent repair deficiency, complementation group 2; General transcription and DNA repair factor IIH helicase subunit XPD; MGC102762; MGC126218; MGC126219; TFIIH; TFIIH 80 kDa subunit; TFIIH basal transcription factor complex 80 kDa subunit; TFIIH basal transcription factor complex helicase subunit; TFIIH basal transcription factor complex helicase XPB subunit; TFIIH basal transcription factor complex helicase XPD subunit; TFIIH p80; TFIIH subunit XPD; TTD; TTD1; xeroderma pigmentosum complementary group D; xeroderma pigmentosum group D-complementing protein; XPD; XPDC | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13871, 2068, 308415 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.69 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O08811, P18074 | |
| ERCC2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human XPD/XPD/ERCC2 (NP_0011243391). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.