missing translation for 'onlineSavingsMsg'
Learn More

XMD8-87, MedChemExpress

Product Code. 16436848
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16436848 10 mg 10mg
16486946 100 mg 100mg
16479387 5 mg 5mg
16443496 25 mg 25mg
16432917 50 mg 50mg
5 options
This item is not returnable. View return policy

Product Code. 16436848

Brand: Medchem Express HY15811

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress XMD8-87 is a potent TNK2 inhibitor with IC50 values of 38 and 113 nM for the D163E and R806Q mutations, respectively.

XMD8-87 is a potent TNK2 inhibitor with IC50 values of 38 and 113 nM for the D163E and R806Q mutations, respectively.

Specifications

Chemical Name or Material XMD8-87
CAS 1234480-46-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H27N7O2
Quantity 10 mg
Synonym ACK1-B19
Solubility Information DMSO : ≥ 26 mg/mL (58.36 mM)
SMILES O=C1C2=CC=CC=C2N(C)C3=NC(NC4=CC=C(N5CCN(C)CC5)C=C4OC)=NC=C3N1
Molecular Weight (g/mol) 445.52
Formula Weight 445.52
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.