Learn More
Tocris Bioscience™ XL 184
Potent VEGFR inhibitor; also inhibits other RTKs
Brand: Tocris Bioscience™ 5422/50
This item is not returnable.
View return policy
Description
XL 184 is a potent VEGFR inhibitor (IC50 = 0.035 nM); also inhibits c-Met, KIT, RET, FLT4, AXL, FLT3, FLT1 and Tie2 (IC50 values are 1.3, 4.6, 5.2, 6, 7, 11.3, 12 and 14.3 nM, respectively). Induces intratumoral hypoxia and apoptosis. Reduces tumor invasion and metastasis in vivo. Antiangiogenic.Specifications
| >98% | |
| Cabozantinib, BMS 907351 | |
| 50 mg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only