Learn More
Tocris Bioscience™ XL 184
GREENER_CHOICE
Potent VEGFR inhibitor; also inhibits other RTKs
Brand:  Tocris Bioscience™ 5422/50
Encompass
Product Code. 15776816
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
XL 184 is a potent VEGFR inhibitor (IC50 = 0.035 nM); also inhibits c-Met, KIT, RET, FLT4, AXL, FLT3, FLT1 and Tie2 (IC50 values are 1.3, 4.6, 5.2, 6, 7, 11.3, 12 and 14.3 nM, respectively). Induces intratumoral hypoxia and apoptosis. Reduces tumor invasion and metastasis in vivo. Antiangiogenic.
Specifications
Specifications
>98%
Cabozantinib, BMS 907351