Learn More
Uvex Xenova™ Black/Blue PU 95567 Boot - Width 10
Uvex 2 Xenova™ design provides comfortable, metal-free safety shoes; light weight with optimum cushioning and breathability for indoor and outdoor work
1445.00 NOK
Specifications
Color | Black, Blue |
---|---|
Material | Leather |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17288451
|
Uvex
9556738 |
38 |
1445.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17298451
|
Uvex
9556739 |
39 |
1445.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17208461
|
Uvex
9556740 |
40 |
1445.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17218461
|
Uvex
9556741 |
41 |
1445.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17228461
|
Uvex
9556742 |
42 |
1445.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17238461
|
Uvex
9556743 |
43 |
1445.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17248461
|
Uvex
9556744 |
44 |
1445.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17258461
|
Uvex
9556745 |
45 |
1445.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17268461
|
Uvex
9556746 |
46 |
1445.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17278461
|
Uvex
9556747 |
47 |
1445.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17288461
|
Uvex
9556748 |
48 |
1445.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17298461
|
Uvex
9556749 |
49 |
1445.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17208471
|
Uvex
9556750 |
50 |
1445.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17218471
|
Uvex
9556751 |
51 |
1445.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17228471
|
Uvex
9556752 |
52 |
1445.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
Uvex 2 Xenova™ Boot 95567 S2 SRC
- Extra wide sports fit
- Latest generation of 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good side stability, no thermal conductivity and anti-magnetic properties
- Breathable, water repellent full grain leather
- Distance mesh lining provides optimum breathability
- Uvex anti-twist heel counter for more stability and prevention of twisting ankle
- Soft padded collar and dust tongue
- Comfortable removable insole
- Antistatic with moisture control system
- Double density PUR sole, non-marking with excellent shock absorption and abrasion resistant outsole
- Self-cleaning tread design
- Oil and fuel resistant
- Free of lacquer wetting inhibitors such as silicones, softeners etc.
- Short term heat resistant up to 120°C
- Antistatic
- Fulfils ESD requirements: resistance to ground Kleiner 35 megohms
- Article No. 95567
- Standard EN ISO 20345:2011 S2 SRC
- Sizes: 38 to 52
Specifications
Black, Blue | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Leather | |
Composite | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S2 |