Learn More
Uvex Xenova™ Black/Blue PU 95563 Boot - Width 12
Uvex 2 Xenova™ design provides comfortable, metal-free safety shoes; light weight with optimum cushioning and breathability for indoor and outdoor work
1485.00 NOK
Specifications
Color | Black, Blue |
---|---|
Material | Leather |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17238441
|
Uvex
9556338 |
38 |
1485.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17248441
|
Uvex
9556339 |
39 |
1485.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17258441
|
Uvex
9556340 |
40 |
1485.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17268441
|
Uvex
9556341 |
41 |
1485.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17278441
|
Uvex
9556342 |
42 |
1485.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17288441
|
Uvex
9556343 |
43 |
1485.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17298441
|
Uvex
9556344 |
44 |
1485.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17208451
|
Uvex
9556345 |
45 |
1485.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17218451
|
Uvex
9556346 |
46 |
1485.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17228451
|
Uvex
9556347 |
47 |
1485.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17238451
|
Uvex
9556348 |
48 |
1485.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17248451
|
Uvex
9556349 |
49 |
1485.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17258451
|
Uvex
9556350 |
50 |
1485.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17268451
|
Uvex
9556351 |
51 |
1485.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
17278451
|
Uvex
9556352 |
52 |
1485.00 NOK
1 pair |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
Uvex 2 Xenova™ Boot 95563 S3 SRC
- Extra wide sports fit
- Latest generation of 100% metal-free Uvex Xenova™ toecap
- Compact design, anatomical shape, good side stability, no thermal conductivity and anti-magnetic properties
- Penetration-resistant, non-metallic midsole in accordance with latest safety standards, does not restrict flexibility
- Breathable water repellent full grain leather
- Scratch resistant scuffcap made from PU coated leather for longer service life
- Distance mesh lining provides optimum breathability
- Uvex anti-twist heel counter for more stability and prevention of twisting ankle
- Soft padded collar and tongue
- Comfortable removable insole, antistatic with moisture control system
- Double density PUR sole, non-marking with excellent shock absorption and abrasion resistant outsole
- Self-cleaning tread design
- Oil and fuel resistant
- Free of lacquer wetting inhibitors such as silicones, softeners, etc.
- Short term heat resistance up to 120°C
- Antistatic
- Fulfills ESD requirements: resistance to ground Kleiner 35 megohms
- Article No. 95563
- Standard EN ISO 20345:2011 S3 SRC
- Sizes: 38 to 52
Specifications
Black, Blue | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Leather | |
Composite | |
EN ISO 20345:2011 | |
Wide width | |
Yes | |
S3 |