Learn More
Trajan Scientific and Medical™ XCHANGE™ CTC PAL-XT Syringe for LC
Product Code. 15259273
Change view
Click to view available options
Volume (Metric):
100 μL
250 μL
500 μL
1 mL
Diameter (Metric) Inner:
0.17 mm
0.375 mm
Unit Size:
Each
Product Code. Volume (Metric) Diameter (Metric) Inner unitSize
15259273 500 μL 0.375 mm Each
15279273 1 mL 0.375 mm Each
15209273 100 μL 0.17 mm Each
15239273 250 μL 0.375 mm Each
4 options
Product Code. 15259273
Brand: Trajan Scientific and Medical™ 2928730
The knowledge and expertise that goes into manufacturing an SGE syringe gives outstanding performance and superior quality
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
Specifications
Description CTC PAL-XT Syringe for LC
Needle Gauge 22 G
Diameter (Metric) Inner Needle 0.375 mm
Diameter (Metric) Inner 0.375 mm
Diameter (Metric) Outer 0.71 mm
Diameter (Metric) Outer Needle 0.71 mm
For Use With (Application) For liquid handling and LC
Volume (Metric) 500 μL
Length (Metric) Needle 56 mm
Needle Point Style Cone