Learn More
XBP1S-Specific Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Product Code. 16883365 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16883365 20 μL 20µL
16873365 150 μL 150µL
2 options
Product Code. 16883365
Brand: Proteintech 248681AP20UL
Rabbit Polyclonal Antibody
The XBP1S-specific antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human XBP1S-specific. This antibody recognizes human, mouse antigen. The XBP1S-specific antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Antigen XBP1S-Specific
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 293 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 7494
Target Species Human, Mouse
Isotype IgG