Learn More
XBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25997-1-AP-20UL
 View more versions of this product
Product Code. 16844525
  • 1710.33 NOK / 20µL
Description
XBP1 encodes a transcription factor that regulates MHC class II genes by binding to a promoter element referred to as an X box. This gene product is a bZIP protein, which was also identified as a cellular transcription factor that binds to an enhancer in the promoter of the T cell leukemia virus type 1 promoter. It may increase expression of viral proteins by acting as the DNA binding partner of a viral transactivator. It has been found that upon accumulation of unfolded proteins in the endoplasmic reticulum (ER), the mRNA of this gene is processed to an active form by an unconventional splicing mechanism that is mediated by the endonuclease inositol-requiring enzyme 1 (IRE1). The resulting loss of 26 nt from the spliced mRNA causes a frame-shift and an isoform XBP1(S), which is the functionally active transcription factor. The isoform encoded by the unspliced mRNA, XBP1(U), is constitutively expressed, and thought to function as a negative feedback regulator of XBP1(S), which shuts off transcription of target genes during the recovery phase of ER stress. A pseudogene of XBP1 has been identified and localized to chromosome 5.
Specifications
Specifications
XBP1
Polyclonal
Unconjugated
XBP1
TREB5, X box binding protein 1, XBP 1, XBP1, XBP2
Rabbit
Antigen Affinity Chromatography
RUO
22433, 289754, 7494
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O35426, P17861, Q9R1S4
Xbp1
XBP1 Fusion Protein Ag21714
20 μL
Primary
Rat, Mouse, Human