missing translation for 'onlineSavingsMsg'
Learn More

XAV-939, MedChemExpress

Product Code. 16441166
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16441166

Brand: Medchem Express HY15147

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress XAV-939 is a potent tankyrase inhibitor that targets Wnt/β-catenin signaling. XAV-939 stabilizes axin by inhibiting tankyrase 1 and tankyrase 2 (IC50s of 5 and 2 nM, respectively), thereby stimulating β-catenin degradation. XAV939 binds tightly to the catalytic (PARP) domains of TNKS1 and TNKS2 (Kds of 99 and 93 nM, respectively).

XAV-939 is a potent tankyrase inhibitor that targets Wnt/β-catenin signaling. XAV-939 stabilizes axin by inhibiting tankyrase 1 and tankyrase 2 (IC50s of 5 and 2 nM, respectively), thereby stimulating β-catenin degradation. XAV939 binds tightly to the catalytic (PARP) domains of TNKS1 and TNKS2 (Kds of 99 and 93 nM, respectively).

Specifications

Chemical Name or Material XAV-939
Physical Form Solid
CAS 284028-89-3
Color Off-White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C14H11F3N2OS
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 25 mg/mL (80.05 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1C(CSCC2)=C2N=C(C3=CC=C(C(F)(F)F)C=C3)N1
Molecular Weight (g/mol) 312.31
Formula Weight 312.31
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.