Learn More
Seward X500 Saco Standard 400ml
Brand:  Seward BA6041/5/500
X500 Saco Standard 400ml
Product Code. 15687338
  • ONLINE EXCLUSIVE PRICE
    1295.00 NOK
    1271.86 NOK / Pack of 500
    Save 23.14 NOK
    1% Off
Only null left
Estimated Shipment: 11-02-2026