Learn More
Eppendorf X500 Protein LoBind Tubes, 2,0ml, PCR clean, 500 pcs.
GREENER_CHOICE
Brand:  Eppendorf 0030108450
1036.90 NOK valid until 2025-12-20
BEST PRICE on Promo! Use promo code "24125" to get your promotional price.
X500 Protein LoBind Tubes, 2,0ml, PCR clean, 500 pcs.
Product Code. 17144364
  • ONLINE EXCLUSIVE PRICE
    1165.00 NOK
    1060.00 NOK / Pack of 500
    Save 105.00 NOK
    9% Off
View all promotions
Only null left
Estimated Shipment: 22-01-2026