Learn More
Kartell X4 Graduated Beaker, low form, PP, 2l, class B, bl
BIOPHARMA
Brand:  Kartell 0182700
476.90 NOK valid until 2025-12-20
BEST PRICE on Promo! Use promo code "24125" to get your promotional price.
X4 Graduated Beaker, low form, PP, 2l, class B, bl
Product Code. 15865567
  • 502.00 NOK / Pack of 4
Only null left
Estimated Shipment: 28-01-2026