Learn More
Bohlender X3 Triangular Magnetic Stirring Bars L 40 mm 14mm
Brand:  Bohlender C357-18
X3 Triangular Magnetic Stirring Bars L 40 mm 14mm
Product Code. 17157034
  • ONLINE EXCLUSIVE PRICE
    308.70 NOK
    240.88 NOK / Pack of 3
    Save 67.82 NOK
    21% Off
Only null left
Estimated Shipment: 02-03-2026