Learn More
Macherey-Nagel™ Cellulose Extraction Thimbles
Minimizes the hands-on time involved in extraction. Macherey Nagel™ Cellulose Extraction Thimbles are used for holding solid materials from which substances are to be extracted with a suitable solvent.
Brand:  Macherey-Nagel™ 645040
Product Code. 10410112
  • ONLINE EXCLUSIVE PRICE
    3845.00 NOK
    3685.12 NOK / Pack of 25
    Save 159.88 NOK
    4% Off
Only null left
Estimated Shipment: 05-02-2026
Description
  • Manufactured from pure cellulose
  • Used in the fields of air and waste gas analysis for collecting solid particles
  • The choice of analytical chemists using Soxhlet or automated extractors
Specifications
Specifications
25 Pk.
Cellulose
275 mm
55 mm
Analysis of soil contaminants, Analysis of soil contaminants
Extraction Thimbles