Learn More
Ansell BioClean-D™ Sterile cleanroom Coverall with Hood and integrated Boots
Shop All Ansell ProductsDescription
- Reduced contamination risks: Lightweight CleanTough™ material means the BioClean-D Coverall with Hood and Integrated Boots – Sterile S-BDFC is low-linting and ISO cleanroom-compliant, reducing contamination risks
- Optimized protection: This sterile coverall features a front zip with a protective flap, resisting liquid splashes and chemicals*, while thumb loops and an elasticated hood, back, cuffs and ankles ensure a secure fit
- Enhanced features: This antistatic coverall features integrated boots and slip-resistant soles, for added workplace safety in laboratory settings
- Sterility assurance level: Special packaging eliminates cross contamination, while gamma irradiation sterilization ensures a minimum SAL of 10-6
- Contact customer service for chemotherapy drug permeation times and recommendations.
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | X-Small |
| Gender | Unisex |
| Material | Flashspun Polyethylene |
| Color | White |
| Disposable | Single-use |
| Waist Style | Standard |
| Ankle Style | Attached Boots, Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |