Learn More
Kartell X12 measuring scoop 50ml PP 27
Brand:  Kartell 0105900
X12 measuring scoop 50ml PP
Product Code. 10376301
  • ONLINE EXCLUSIVE PRICE
    335.00 NOK
    311.65 NOK / Pack of 12
    Save 23.35 NOK
    6% Off
Only null left
Estimated Shipment: 19-01-2026