Learn More
Epredia X1000 Round Coverslip dia. 5mm #1 (0.13-0.16mm)
Brand:  Epredia CB00050RA120MNZ0
X1000 Round Coverslip dia. 5mm #1 (0.13-0.16mm)
Product Code. 11888372
  • ONLINE EXCLUSIVE PRICE
    752.00 NOK
    741.17 NOK / Pack of 1000
    Save 10.83 NOK
    1% Off
 Restricted Item
Estimated Shipment: 05-01-2027