Learn More
Epredia X1000 Round Coverslip dia. 18mm #1 (0.13-0.16mm)
Brand:  Epredia CB00180RA120MNZ0
592.75 NOK valid until 2026-03-31
BEST PRICE on Promo! Use promo code "27994" to get your promotional price.
X1000 Round Coverslip dia. 18mm #1 (0.13-0.16mm)
Product Code. 11709875
  • 797.00 NOK / Pack of 1000
Save Big on Labware
Use promo code 27994
View all promotions
Only null left
Estimated Shipment: 09-02-2026