Learn More
Epredia X1000 Round Coverslip dia. 12mm #0 (0.08-0.12mm)
Brand:  Epredia CB00120RA020MNZ0
762.50 NOK valid until 2026-03-31
BEST PRICE on Promo! Use promo code "27994" to get your promotional price.
X1000 Round Coverslip dia. 12mm #0 (0.08-0.12mm)
Product Code. 11856933
  • 852.00 NOK / Pack of 1000
Save Big on Labware
Use promo code 27994
View all promotions
Only null left
Estimated Shipment: 18-02-2026