Learn More
Sampling systems X10 SteriWare Funnel 200mm
BIOPHARMA
Brand:  Sampling systems 8370H-200
X10 SteriWare Funnel 200mm
Product Code. 17838268
  • ONLINE EXCLUSIVE PRICE
    534.00 NOK
    449.29 NOK / Pack of 10
    Save 84.71 NOK
    15% Off
Only null left
Estimated Shipment: 04-03-2026