Learn More
Trajan X10 1/8 in. GC Ferrules 0.5 mm ID Two Hole Vespel
Brand:  Trajan 212222
X10 1/8 in. GC Ferrules 0.5 mm ID Two Hole Vespel
Product Code. 17842388
  • 1015.00 NOK / Pack of 10
Only null left
Estimated Shipment: 23-06-2026