Learn More
Corning™ X-SERIES™ Balance Ring Kit
Product Code. 17224466 Shop All Corning Products
Change view
Click to view available options
Quantity:
1 set
Unit Size:
1 set
Product Code. Quantity unitSize
17224466 1 set 1 set
1 options
Product Code. 17224466
Brand: Corning™ 6923
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Corning™ X-SERIES™ Balance Ring Kit is designed for balancing contents of centrifuge cup prior to centrifugation.
The Corning™ X-SERIES™ Systems are manufactured for Corning™ by ThermoGenesis Corp.
X-SERIES™, X-WASH™, X-LAB™, X-BACS™, and X-MINI™ are either registered trademarks or trademarks of ThermoGenesis Corp. in the United States and/or other countries.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 2.5 and 7.4 g
For Use With (Equipment) X-WASH and X-LAB Systems
Quantity 1 set