Learn More
ABEBA™ X-Light White Occupational Clogs OB
Description
- Lightweight, comfort, hygiene
- Comply to EN ISO 20347 : 2012, OB SRC
- ESD dissipative according to EN 61340 Climate class 1
- Slip resistant Mono PU outsole SRC
- Smooth leather
- Silver ions lining
- Fixed heel strap
- Replaceable Active Comfort insole
- Sanitized treatment
Specifications
Specifications
| Certifications/Compliance | CE EN ISO 20347:2012 OB, FO, SRC, ESD |
| Color | White |
| Liner Material | Breathable lining with Silver ions |
| Material | Leather |
| Size | 35 |
| Sole Features | Slip and abrasion resistant, shock absorbing, ESD dissipative |
| Sole Material | Polyurethane |
| Style | Clogs |
| Packaging Quantity | 1 Pair |
| Certifications/Compliance (Europe) | CE EN ISO 20347:2012, DIN EN 61340 |
| Show More |