Learn More
Abeba™ X-Light Low Safety Shoe
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Description
- Size 35 to 48
- CE, EN ISO 20345:2011, S2, SRC
- Smooth leather, black
- Breathable lining with Silver ions
- Instep area with lacing
- Slip resistance PU outsole sole
- Steel toe cap
- Replaceable Active Comfort Insole (art. 351620)
- Antistatic
- Reflective material
Specifications
Specifications
| Closure Type | Laces |
| Color | Black |
| Material | Leather |
| Size | 39 |
| Toe Material | Steel |
| Certifications/Compliance (Europe) | CE, EN ISO 20345:2011, S2, SRC |