Learn More
Abeba™ X-Light Low Safety Shoe
Product Code. 17503165
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
17503165 40 1 pair
17553155 35 1 pair
17563155 36 1 pair
17573155 37 1 pair
17583155 38 1 pair
17593155 39 1 pair
17513165 41 1 pair
17523165 42 1 pair
17533165 43 1 pair
17543165 44 1 pair
17553165 45 1 pair
17563165 46 1 pair
17573165 47 1 pair
17583165 48 1 pair
14 options
Product Code. 17503165
Brand: Abeba™ 711038/40
Only null left
Add to basket
Add to basket
Abeba X-Light Low Safety Shoe has a breathable inner lining with silver ions.
  • Size 35 to 48
  • CE, EN ISO 20345:2011, S2, SRC
  • Smooth leather, black
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Slip resistance PU outsole sole
  • Steel toe cap
  • Replaceable Active Comfort Insole (art. 351620)
  • Antistatic
  • Reflective material
Specifications
Closure Type Laces
Color Black
Material Leather
Size 40
Toe Material Steel
Certifications/Compliance (Europe) CE, EN ISO 20345:2011, S2, SRC