Learn More
ABEBA™ X-Light Black Safety Trainers S3 ESD
Product Code. 17101585
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17101585 35 1 pair
17111585 36 1 pair
17101595 45 1 pair
17111595 46 1 pair
17121595 47 1 pair
17131595 48 1 pair
17121585 37 1 pair
17131585 38 1 pair
17141585 39 1 pair
17151585 40 1 pair
17161585 41 1 pair
17171585 42 1 pair
17181585 43 1 pair
17191585 44 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 17101585 Supplier ABEBA™ Supplier No. 7131874/35
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Black X-light safety trainer with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, textile antipuncture midsole, PU slip resistant outsole
  • Lightweight, comfortable safety trainer
  • Comply to EN ISO 20345: 2012, S3 SRC
  • ESD dissipative according EN 61340 class climatic 1
  • Slip resistant Mono PU outsole SRC
  • Steel toe cap (200 J)
  • Textile antipuncture midsole
  • Black smooth leather
  • Silver ions lining
  • Replaceable active comfort insole
Specifications
Color Black
Material Leather
Size 35
Toe Material Steel
Certifications/Compliance (Europe) Comply to EN 20345: 2012 S3 SRC, ESD according EN 61340
Gender Unisex
Ankle Style Low-cut
Waterproof Water Resistant
Product Type S3 SRC ESD