Learn More
ABEBA™ X-Light Black Safety Clog Shoes SB
Product Code. 16338331
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 16338331
Brand: ABEBA™ 7131035/47
Only null left
Add to basket
Add to basket
ABEBA™ X-Light Black Safety Clog Shoes SB with silver ions lining to reduce odor, replaceable Active Comfort insole, and a fixed heel strap.
  • Lightweight, comfort, hygiene
  • Comply to EN ISO 20345 : 2012, S2 SRC
  • ESD dissipative according to EN 61340 Climate class 1
  • Slip resistant Mono PU outsole SRC
  • Steel toe cap (200 joules)
  • Black smooth leather
  • Silver ions lining
  • Replaceable Active Comfort insole
  • Sanitized treatment
Specifications
Buckle Material Plastic
Certifications/Compliance CE EN ISO 20345:2012 SB, FO, SRC, ESD
Closure Type Heel strap non-foldable, adjustable
Color Black
Liner Material Breathable lining with Silver ions
Material Leather
Size 47
Sole Features Slip and abrasion resistant, shock absorbing, ESD dissipative
Sole Material Polyurethane
Style Clogs
Toe Material Steel
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) CE EN ISO 20345:2012, DIN EN 61340
Resistant To Water