Learn More
ABEBA™ X-Light Black Occupational Clogs OB
Product Code. 16368391
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 16368391
Brand: ABEBA™ 7131135/40
Only null left
Add to basket
Add to basket
ABEBA™ X-Light Black Occupational Clogs OB with silver ions lining to reduce odor, a replaceable Active Comfort insole, and a non-foldable, adjustable heel strap.
  • Lightweight, comfort, hygiene
  • Comply to EN ISO 20347 : 2012, OB SRC
  • ESD dissipative according to EN 61340 Climate class 1
  • Slip resistant Mono PU outsole SRC
  • Smooth leather
  • Silver ions lining
  • Heel strap non-foldable, adjustable
  • Replaceable Active Comfort insole
  • Sanitized treatment
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB, FO, SRC, ESD
Color Black
Liner Material Breathable lining with Silver ions
Material Leather
Size 40
Sole Features Slip and abrasion resistant, shock absorbing, ESD dissipative
Sole Material Polyurethane
Style Clogs
Packaging Quantity 1 Pair
Certifications/Compliance (Europe) CE EN ISO 20347:2012, DIN EN 61340
Resistant To Water