Learn More
Tocris Bioscience™ WZ 4003
GREENER_CHOICE
Potent and selective NUAK1/2 inhibitor
Brand:  Tocris Bioscience™ 5177/50
Encompass
Product Code. 15726546
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
WZ 4003 is a potent and selective NUAK1/2 inhibitor (IC50 values are 20 and 100 nM respectively). Exhibits no significant inhibition against a panel of 139 kinases, including ten AMPK family members. Inhibits NUAK1-mediated MYPT1 phosphorylation. Also inhibits cell proliferation in U2OS cells.
Specifications
Specifications
>98%