Learn More
Tocris Bioscience™ WZ 4003
Potent and selective NUAK1/2 inhibitor
Brand: Tocris Bioscience™ 5177/50
This item is not returnable.
View return policy
Description
WZ 4003 is a potent and selective NUAK1/2 inhibitor (IC50 values are 20 and 100 nM respectively). Exhibits no significant inhibition against a panel of 139 kinases, including ten AMPK family members. Inhibits NUAK1-mediated MYPT1 phosphorylation. Also inhibits cell proliferation in U2OS cells.Specifications
| >98% | |
| 50 mg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only