Learn More
Kimberly-Clark™ WypAll™ X60 General Clean™ Quarterfold Reusable Cleaning Cloths 6034, 12 Packs x 76
Product Code. 10499272
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 600
Product Code. Color unitSize
10499272 White Pack of 600
1 options
Product Code. 10499272
Brand: Kimberly-Clark™ 6034
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Reusable quarter-fold cleaning cloths are versatile and designed for light duty and general wiping tasks. Supplied in a protective bag and feature advanced Hydroknit™ technology for a lightweight, absorbent cleaning cloth. Quantity 12 packs x 76 cloths (912 Total).
  • Each WypAll X60 General Clean™ quarter-fold cleaning cloths contain 76 white cloths
  • HyperAbsorb™ technology helps reduce waste and drive sustainable practices by providing great durability and allowing reuse up to 5 times
  • Advanced Hydroknit™ technology ensures these professional cleaning cloths are strong and fast absorbing, hydroentangling process bonds cellulose fibres into webs for faster absorption and greater tear resistance
  • Reusable cleaning cloths have been tested as compatible for use in spray and wipe applications with peracetic acid, hydrogen peroxide, bleach (sodium hypochlorite) and quats (quaternary ammonium compounds)
  • Always follow manufacturers' instructions for dilution, application, and contact time
  • High-capacity cleaning cloths roll, ideal for larger manufacturing and industrial work areas, or anywhere requiring extensive wiping
  • Compatible with compatible with WypAll Wiper Dispenser (product code 7969)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Non-woven Wipers
Length (English) 11.02 in.
Length (Metric) 50 cm
Width (English) 11.02 in
Width (Metric) 28.4 cm
Material Polypropylene
Basis Weight 5.59kg
Color White
Format 1/4 fold
For Use With (Application) Light-duty clean-up
Packaging Quantity 12 Bags x 76 Sheets
Ply 1-ply