Learn More
Kimberly-Clark™ WypAll™ L20 General Clean™ Medium Duty Wipes, 2 Ply Centrefeed Roll, 6 Rolls x 380 Sheets
Product Code. 15656485
Change view
Click to view available options
Color:
Blue
White
Unit Size:
6 rolls
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15656485 Blue 6 rolls
15666485 White 6 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15656485 Supplier Kimberly-Clark™ Supplier No. 7302
  • ONLINE EXCLUSIVE PRICE
    1490.00 NOK
    628.45 NOK / 6 rolls
    Save 861.55 NOK
    57% Off
View all promotions
Estimated Shipment: 22-12-2026
Single-use 2-ply wiping paper is designed for heavy-duty wiping tasks. Unique Airflex™ technology delivers exceptional strength and absorbency. Compatible with Aquarius™ dispenser 7017. Quantity 6 rolls x 380 sheets, 2 ply (2,280 total)
  • Each individual roll contains 380, 2 ply sheets
  • Single-use wiping paper designed for heavy-duty wiping tasks
  • Unique Airflex™ technology delivers exceptional strength and absorbency
  • Centrefeed roll format
  • Compatible with WypAll Centrefeed Wiper Dispenser (product code 7017)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Heavy-duty Wiper
Length (English) 15.35 in.
Length (Metric) 58.5 cm
Width (English) 15.35 in
Width (Metric) 39 cm
Material Paper
Color Blue
Disposable Single-use
Dimensions (L x W) 42.50 x 18.50 cm
For Use With (Application) Highly absorbent single use wipes for multipurpose wiping and cleaning: medium duty wiping tasks such as polishing glass, smooth surfaces and cleaning tools
For Use With (Equipment) Can be used with a dispensing system (7087 or 7088) to help control use and reduce wastage
Packaging Quantity 6 Rolls x 300 Sheets
Ply 2-ply