Learn More
WTW™ WTW™ Rectangular Cuvette
Product Code. 10693372
Change view
Click to view available options
Path Length:
10 mm
20 mm
50 mm
Unit Size:
Each
Product Code. Path Length unitSize
10693372 10 mm Each
10508381 50 mm Each
10324781 20 mm Each
3 options
Product Code. 10693372
Brand: WTW™ 250605
Only null left
Add to basket
Add to basket
For automatic detection of the photLab™ filter and spectral photometer
The glass and quartz cuvettes with matte side surface are suitable for the automatic cuvette detection of the photoLab™ filter and spectral photometer. When using disposable cuvettes, the material PMMA as well as a structured or matte side surface is required due to the cuvette detection by the photoLab™ photometers. The glass and quartz cuvettes have a marking in order to place the cuvettes into the photometer in the same direction for an even higher precision.Cuvettes for turbidity measurement can be marked to prevent influences by glass: This allows a higher accuracy, also for values < 1 NTU.
TRUSTED_SUSTAINABILITY
Specifications
Type Cuvette
Path Length 10 mm
Shape Rectangle