Learn More
WT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12609-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16850414
  • 1415.13 NOK / 20µL
Estimated Shipment: 13-10-2023
Description
Wilm′s Tumor (WT), a sporadic and familial childhood kidney tumor, is genetically heterogeneous. Wilm′s tumor is associated with mutations of WT1, a zinc-finger transcription factor that is essential for the development of the metanephric kidney and the urogenital system. The WT1 gene is normally expressed in fetal kidney and mesothelioma.
Specifications
Specifications
WT1
Polyclonal
Unconjugated
WT1
GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT 2, WT1, WT33
Rabbit
Antigen Affinity Chromatography
RUO
22431, 24883, 7490
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P19544, P22561, P49952
WT1
WT1 Fusion Protein Ag3337
20 μL
Primary
Mouse, Human, Rat