Learn More
Cole Parmer™ Write-On Tape
Product Code. 15719404 Shop All Cole-Parmer Products
Change view
Click to view available options
Color:
Assorted
White
Width (English):
0.5 in.
Unit Size:
Pack of 24
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Width (English) Unit Size
15719404 White 0.5 in. Pack of 6
15366155 Assorted 0.5 in. Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15719404 Supplier Cole Parmer™ Supplier No. 0653021
  • ONLINE EXCLUSIVE PRICE
    607.00 NOK
    193.45 NOK / Pack of 6
    Save 413.55 NOK
    68% Off
Estimated Shipment: 14-01-2027
Use to label/identify laboratory items/products.
  • Identify with writing and color.
  • Durable tape is waterproof, and oil and acid resistant.
  • Can be used with pencil, pen, or permanent marker.
  • Temperature resistant.
Specifications
Color White
No. per Pack 6/Pk.
Quantity 6/Pk.
Temperature Range (Metric) -23°C to +121°C
Dimensions (L x W) 14 yd. x 0.5 in.
Length (English) 14 yd.
Width (English) 0.5 in.