Learn More
Microspec™ Wrappastack™ Sleeves
Product Code. 15702145
Change view
Click to view available options
Diameter (Metric):
55 mm
90 mm
Unit Size:
Pack of 200
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
15702145 55 mm Pack of 200
15772135 90 mm Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15702145 Supplier Microspec™ Supplier No. K5
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
For storage of prepared petri dishes.
  • Professionally wrap poured plates for fridge storage or incubation
  • Allows poured plates to heat shrunk into sleeves for fridge storage
  • Keeps plates neatly stacked and steady
  • Reduces moisture loss
  • Tear strip for easy removal of plates when required
Specifications
Material Polypropylene
Type Sleeve
Height (Metric) 180 mm
Diameter (Metric) 55 mm