Learn More
BD Plastipak™ Wound and Bladder Syringe
Click to view available options
Volume (Metric):
50 mL
Unit Size:
Pack of 60
Description
- Very high tightness due to tight, double-sealing ring
- Retaining ring prevents accidental plunger rod withdrawal
- Centered catheter tip attachment
- Silicone oil lubricant ensures piston slides gradually and evenly
- Spray cylinder and piston rod made of polypropylene (PVC-free)
- Disposable syringes are EO-sterilized and individually packaged
Specifications
Specifications
| Material | Polypropylene |
| Color | Clear |
| Diameter (Metric) Outer | 29.4 mm |
| Latex Free | Yes |
| Length (Metric) | 133 mm |
| Product Type | Wound/Bladder Syringe |
| Disposable | Single-use |
| Graduations | 1 mL |
| Quantity | 60 Pk. |
| Volume (Metric) | 50 mL |
| Show More |