Learn More
Sartorius™ Worktable for Safety Weighing Cabinet SWC working area 1250 x 750 mm
Product Code. 15519734 Shop All Sartorius Products
Change view
Click to view available options
Dimensions (L x W):
1250 x 750 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15519734 1250 x 750 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15519734 Supplier Sartorius™ Supplier No. YWT11
  • ONLINE EXCLUSIVE PRICE
    25470.00 NOK
    23118.93 NOK / Pack of 1
    Save 2351.07 NOK
    9% Off
 Restricted Item
Estimated Shipment: 09-12-2026
For use with Sartorius safety weighing cabinets.
  • Accessory item
  • Lab bench
  • Working area of 1250 x 750 mm
TRUSTED_SUSTAINABILITY
Specifications
Product Type Work Surface
Dimensions (L x W) 1250 x 750 mm
For Use With (Application) Provides working area
For Use With (Equipment) Safety weighing cabinets