Learn More
- Home
- Products
- Facility Safety and Maintenance
- Labels and Label Makers
- Custom Labels
- Brady™ WorkHorse™ B-423 Permanent Polyester Labels
Brady™ WorkHorse™ B-423 Permanent Polyester Labels
Product Code. 11765693| Product Code. | Size (English) | Quantity | unitSize |
|---|---|---|---|
| 11765693 | 1.25 x 0.25 in. (W x H) | 750/Roll | Each |
| 11528614 | 50 ft. x 0.5 in. | 1 Ea. | Each |
| 11751835 | 1 x 0.375 in. (W x H) | 5000/Roll | Each |
| 11721835 | 1 x 0.5 in. (W x H) | 5000/Roll | Each |
| 11731815 | 1 x 1 in. (W x H) | 2500/Roll | Each |
| 11781755 | 2 x 1 in. (W x H) | 1000/Pk. | Each |
| 11761835 | 1.5 x 0.5 in. (W x H) | 5000/Roll | Each |
| 11771835 | 0.5 x 0.5 in. | 5000/Roll | Each |
| 11725693 | 0.9 x 0.25 in. (W x H) | 750/Roll | Each |
| 11745693 | 1 x 0.187 in. (W x H) | 750/Roll | Each |
| 11755693 | 1 x 0.275 in. (W x H) | 750/Roll | Each |
| 11755653 | 3 x 1 in. (W x H) | 100/Pk. | Each |
| 11765653 | 4 x 1 in. (W x H) | 100/Pk. | Each |
| 11777354 | 1.13 x 0.5 in. (W x H) | 500/Roll | Each |
| 11775653 | 2 x 0.25 in. (W x H) | 100/Pk. | Each |
| 11785653 | 2.75 x 1.25 in. (W x H) | 100/Pk. | Each |
| 11775693 | 1.5 x 0.125 in. (W x H) | 750/Roll | Each |
| 11785693 | 1.5 x 0.25 in. (W x H) | 750/Roll | Each |
| 11715683 | 1.5 x 0.5 in. (W x H) | 500/Roll | Each |
| 12890059 | 1.5 x 1.5 in. | 250/Roll | Each |
| 11745683 | 0.375 x 0.375 in. | 500/Roll | Each |
| 11705663 | 3 x 1.9 in. (W x H) | 100/Pk. | Each |
| 11715663 | 4 x 1.9 in. (W x H) | 100/Pk. | Each |
| 11755683 | 0.4 x 0.4 in. (W x H) | 500/Roll | Each |
| 11735693 | 0.25 x 0.25 in. | 750/Roll | Each |
| 11795693 | 0.5 x 0.2 in. (W x H) | 750/Roll | Each |
| 11765683 | 0.5 x 0.5 in. | 500/Roll | Each |
Product Code. 11765693
Brand: Brady™ 114786
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Specifications
| Certifications/Compliance | UL recognized, CSA accepted, AGA approved, Halogen free (DIN VDE 0472/part 815) |
| Description | Abrasion, Chemical, Corrosion, Humidity, UV and Weather-Resistant |
| Adhesive Type | Acrylic, Permanent |
| Color | White |
| Finish | Gloss |
| Material | Polyester (B-423) |
| Product Type | Thermal Transfer Printer Label |
| For Use With (Application) | Asset tracking, Barcoding, Component ID, Equipment ID, General purpose ID, Rating plates |
| For Use With (Equipment) | BMP™71 Printer |
| Labels per Row | 1 |
| Quantity | 750/Roll |
| Shape | Rectangle |
| Size (English) | 1.25 x 0.25 in. (W x H) |
| Size (Metric) | 31.7 x 6.35 mm (W x H) |
| Temperature Range (Metric) | -70°C to +110°C |
| Thickness (English) | 0.003 in. |
| Thickness (Metric) | 0.076 mm |
| Vertical Repeat (Metric) | 9.53 mm |
| Dimensions (LxW) | 6.35 x 31.75 mm |
| Width (Metric) | 31.75 mm |
| Height (Metric) | 6.35 mm |
| Show More Show Less |