Learn More
Heinz Herenz Wooden Sticks
Click to view available options
Tip Type:
Standard
Unit Size:
Pack of 100
Description
- Wooden sticks
- Made of beech and birch
- 150 x 3 mm
- One end pointed
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Sterility | Non-sterile |
| Handle Material | Birch |
| Product Type | Stick |
| Tip Material | Wood |
| Tip Type | Standard |