Learn More
Pinnacle Brush™ Wooden Beaker Brushes
Product Code. 12801934
Change view
Click to view available options
Size:
80 x 50 x 150 mm
Brush Circumference: 18 cm
Unit Size:
Each
Pack of 6
Product Code. Size unitSize
12801934 80 x 50 x 150 mm Each
12891924 Brush Circumference: 18 cm Pack of 6
2 options
Product Code. 12801934
Brand: Pinnacle Brush™ D1225/1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Features center set bristles. Pinnacle Brush™ Tube Wooden Beaker Brushes offer gentle cleaning for beakers.
  • Rounded shape
  • Wood handle
Specifications
Product Type Beaker Brush
Handle Material Wood
For Use With (Equipment) Beaker
Length (Metric) 400 mm
Quantity 1 Ea.
Size 80 x 50 x 150 mm